Learn More
Electron Microscopy Sciences Cotton Tip Applicator Double-End
SDP
Catalog No. 50364973
Encompass_Preferred
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft. The pointed head size is 3/16"(d) tapering to 1/32"(d) x 5/8 "(L). Regular head size 3/16(d) x 5/8 "(L).
Catalog No. 50-364-973 Supplier Electron Microscopy Sciences Supplier No. 7230805
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft. The pointed head size is 3/16"(d) tapering to 1/32"(d) x 5/8 "(L). Regular head size 3/16(d) x 5/8 "(L).