Learn More
Electron Microscopy Sciences Cotton Tip Applicator Large Tip
SDP
Supplier:  Electron Microscopy Sciences 72309
Encompass_Preferred
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft. The pointed head size is 3/16"(d) tapering to 1/32"(d) x 5/8 "(L). Regular head size 3/16(d) x 5/8 "(L). Packed 100 sticks in a sealed bag. Large Tip 15⁄64" Dia. x 11⁄16"L. Wooden Shaft
Catalog No. 50-949-145