Learn More
StatLab Cotton Tip Applicator Stick
SDP
Catalog No. 22000027
Change view
Click to view available options
Tip Type:
Rounded
Catalog No. Tip Type
22-000-027 Rounded
1 options
Catalog No. 22-000-027 Supplier StatLab Supplier No. CTA806W
Add to Cart
Edge
Add to Cart
6 in. non-sterile, splinter-free applicator stick with cotton tip
  • For application of tissue marking dyes and other routine uses in paraffin and frozen sectioning
  • BX1000
Specifications
Sterility Non-sterile
Product Type Applicator Stick
Tip Material Cotton
Autoclavable Not Autoclavable
Handle Material Wood
Tip Type Rounded