Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cottonseed flour, MilliporeSigma™ Supelco™

Catalog No. 111015158 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5158 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5158 Supplier MilliporeSigma Supelco Supplier No. C4898500G
Add to Cart
Add to Cart

Cottonseed flour is a high protein concentrate, which can be obtained from the kernel of the cottonseed after the removal of the oil from the seed.

TRUSTED_SUSTAINABILITY

Specifications

CAS 68308-87-2
Physical Form Neat
MDL Number MFCD00130871
Type Analytical Standard
Quantity 500 g
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.