Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cottonseed Oil, MP Biomedicals

Catalog No. ICN90141901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 lb.
25 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90141901 1 lb.
ICN90141925 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN90141901 Supplier MP Biomedicals, Inc Supplier No. 0290141901
Add to Cart
Add to Cart

Saponification Number: 190-198; Iodine Number: 105-114

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-29-4

Specifications

Quantity 1 lb.
Chemical Name or Material Cottonseed Oil
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.