Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coulomat C, Aquastar™, (Safer Formulation), MilliporeSigma™

Catalog No. MAX1697CN30 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAX1697CN30 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX1697CN30 Supplier MilliporeSigma Supplier No. AX1697CN30

missing translation for 'validMainframeMsg'

For Moisture Analysis

Recommended Storage

Store at room temperature

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Mixture
Synonym Karl Fischer

Specifications

Color Colorless to Yellow
Physical Form Clear Liquid
Quantity 2 L
Packaging Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.