Learn More
Beckman Coulter Coulter Clenz Cleaning Agent, 5 L
Catalog No. 15655120
Change view
Click to view available options
Quantity:
5 L
Catalog No. Quantity
15-655-120 5 L
1 options
This item is not returnable. View return policy
Catalog No. 15-655-120 Supplier Beckman Coulter Supplier No. 8546930
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Coulter Clenz Cleaning Agent is recommended for use with the Multisizer 4e and other Coulter Counters. Clenz Cleaning Agent is packaged in a 5 L cubitainer which requires a spigot to dispense.
  • A cleaning agent that strips protein build-up to aid in preventative maintenance
  • Packaged in a 5 liter container, spigot recommended for pouring (Sold separately)
Specifications
Product Type Cleaning Agent
Capacity (Metric) 5 L
Quantity 5 L
For Use With (Application) Removal of Protein Build-Up
For Use With (Equipment) Multisizer™ 4e Coulter Counters