Learn More
AdipoGen Coumaphos
SDP
Catalog No. 502561372
Encompass_Preferred
Chemical. CAS 56-72-4. Formula C14H16ClO5PS. MW 362.77. Synthetic. An organophosphate pesticide. Acetylcholinesterase AChE inhibitor in the nervous system with subsequent accumulation of toxic levels of acetylcholine ACh. Selective insecticide used for control of a wide variety of livestock insects, including cattle grubs, screw-worms, lice, scabies, flies and ticks. Also used against ectoparasites, which are insects that live on the outside of host animals such as sheep, goats, horses, pigs and poultry. Not used on agricultural or residential plants.
Catalog No. 50-256-1372 Supplier AdipoGen Supplier No. CDXC0350G001
Chemical. CAS 56-72-4. Formula C14H16ClO5PS. MW 362.77. Synthetic. An organophosphate pesticide. Acetylcholinesterase AChE inhibitor in the nervous system with subsequent accumulation of toxic levels of acetylcholine ACh. Selective insecticide used for control of a wide variety of livestock insects, including cattle grubs, screw-worms, lice, scabies, flies and ticks. Also used against ectoparasites, which are insects that live on the outside of host animals such as sheep, goats, horses, pigs and poultry. Not used on agricultural or residential plants.