Learn More
AdipoGen Coumarin 102
SDP
Catalog No. 502561472
Encompass_Preferred
Chemical. CAS 41267-76-9. Formula C16H17NO2. Molecular Weight 255.31. Coumarin 102 also known as coumarin 480, is a laser dye and is widely used as a dynamic fluorescent probe of both small molecules and host cavities, micelles, polymers and solids. Coumarin 102 belongs to 7-aminocoumarins with a structurally rigidized amino group. Spectral data lambdaex 390nm, lambdaem 466nm in ethanol, lambdalaser 480nm 461-513nm range.
Catalog No. 50-256-1472 Supplier AdipoGen Supplier No. CDXC0096M250
Chemical. CAS 41267-76-9. Formula C16H17NO2. Molecular Weight 255.31. Coumarin 102 also known as coumarin 480, is a laser dye and is widely used as a dynamic fluorescent probe of both small molecules and host cavities, micelles, polymers and solids. Coumarin 102 belongs to 7-aminocoumarins with a structurally rigidized amino group. Spectral data lambdaex 390nm, lambdaem 466nm in ethanol, lambdalaser 480nm 461-513nm range.