Learn More
Medchemexpress LLC Coumarin 106 | 41175-45-5 | MFCD00041949 | 6498.0% | 281.35 g/mol | C18H19NO2 | 10 MG
SDP
Catalog No. 5000282056
Encompass_Preferred
Coumarin 106 is a dipolar laser dye with fluorescent properties that also displays cholinesterase inhibitory activity. Supplied as a research-grade solid, it is used in fluorescence studies, dye-laser applications, and biochemical assays where cholinesterase modulation or fluorescent probes are required.
  • Dipolar laser dye with fluorescent properties.
  • Demonstrates AChE and BChE inhibition for biochemical studies.
  • Molecular formula C18H19NO2; molecular weight 281.35 g/mol.
  • Available in small research pack sizes suitable for analytical use.
  • Supplied as a solid for standard laboratory storage and handling.
Catalog No. 50-002-82056 Supplier Medchemexpress LLC Supplier No. HYW32439110MG
Coumarin 106 is a dipolar laser dye with fluorescent properties that also displays cholinesterase inhibitory activity. Supplied as a research-grade solid, it is used in fluorescence studies, dye-laser applications, and biochemical assays where cholinesterase modulation or fluorescent probes are required.
  • Dipolar laser dye with fluorescent properties.
  • Demonstrates AChE and BChE inhibition for biochemical studies.
  • Molecular formula C18H19NO2; molecular weight 281.35 g/mol.
  • Available in small research pack sizes suitable for analytical use.
  • Supplied as a solid for standard laboratory storage and handling.