Learn More
Medchemexpress LLC Coumarin 106 | 41175-45-5 | MFCD00041949 | >98.0% | 281.35 | C18H19NO2 | 50 MG
SDP
Catalog No. 5000281972
Encompass_Preferred
Coumarin 106 is a dipolar laser dye used in research that also functions as a cholinesterase inhibitor. It exhibits mixed-type inhibition of acetylcholinesterase (reported pIC50 4.97, Ki 2.36 μM) and lower potency against butyrylcholinesterase (pIC50 4.56). Molecular formula C18H19NO2 and molecular weight 281.35. In vitro data report no toxicity in Caco-2 cells.
  • Dipolar laser dye useful for fluorescence and photonic applications.
  • Inhibits acetylcholinesterase with reported pIC50 4.97 and Ki 2.36 μM.
  • Also inhibits butyrylcholinesterase with pIC50 4.56.
  • Molecular weight 281.35 and formula C18H19NO2.
  • Reported non-toxic in Caco-2 cell assays.
  • Supplied as a 50 mg research reagent.
Catalog No. 50-002-81972 Supplier Medchemexpress LLC Supplier No. HYW32439150MG
Coumarin 106 is a dipolar laser dye used in research that also functions as a cholinesterase inhibitor. It exhibits mixed-type inhibition of acetylcholinesterase (reported pIC50 4.97, Ki 2.36 μM) and lower potency against butyrylcholinesterase (pIC50 4.56). Molecular formula C18H19NO2 and molecular weight 281.35. In vitro data report no toxicity in Caco-2 cells.
  • Dipolar laser dye useful for fluorescence and photonic applications.
  • Inhibits acetylcholinesterase with reported pIC50 4.97 and Ki 2.36 μM.
  • Also inhibits butyrylcholinesterase with pIC50 4.56.
  • Molecular weight 281.35 and formula C18H19NO2.
  • Reported non-toxic in Caco-2 cell assays.
  • Supplied as a 50 mg research reagent.