Learn More
Medchemexpress LLC Coumarin 106 | 41175-45-5 | MFCD00041949 | >98.0% | 281.35 g/mol | C18H19NO2 | 25 MG
SDP
Catalog No. 5000282177
Encompass_Preferred
Coumarin 106 is a dipolar laser dye used as a research reagent with reported acetylcholinesterase and butyrylcholinesterase inhibitory activity. The compound has molecular formula C18H19NO2, molecular weight 281.35 g/mol, and CAS number 41175-45-5. It is commonly used in photophysical studies and enzymology research and is soluble in DMSO at concentrations ≥25 mg/mL.
Catalog No. 50-002-82177 Supplier Medchemexpress LLC Supplier No. HYW32439125MG
Coumarin 106 is a dipolar laser dye used as a research reagent with reported acetylcholinesterase and butyrylcholinesterase inhibitory activity. The compound has molecular formula C18H19NO2, molecular weight 281.35 g/mol, and CAS number 41175-45-5. It is commonly used in photophysical studies and enzymology research and is soluble in DMSO at concentrations ≥25 mg/mL.