Learn More
AdipoGen Coumarin 153
SDP
Catalog No. 502561509
Encompass_Preferred
Chemical. CAS 53518-18-6. Formula C16H14NO2F3. Molecular Weight 309.28. Coumarin 153 is a well known polar laser dye. Spectral data lambdaem 532nm in ethanol Lasing peak 550nm, Lasing range 521-605nm. It can also be used as green phosphorescent dopant material for organic light-emitting diodes OLED.
Catalog No. 50-256-1509 Supplier AdipoGen Supplier No. CDXC0054G001
Chemical. CAS 53518-18-6. Formula C16H14NO2F3. Molecular Weight 309.28. Coumarin 153 is a well known polar laser dye. Spectral data lambdaem 532nm in ethanol Lasing peak 550nm, Lasing range 521-605nm. It can also be used as green phosphorescent dopant material for organic light-emitting diodes OLED.