Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 2, 99%, laser grade

Catalog No. ac405485000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC405485000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC405485000 Supplier Thermo Scientific Chemicals Supplier No. 405485000
Only null left
Add to Cart

CAS: 26078-25-1 | C13H15NO2 | 217.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26078-25-1
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.268
MDL Number MFCD00006860
InChI Key QZXAEJGHNXJTSE-UHFFFAOYSA-N
Synonym 4, 6-Dimethyl-7-ethylaminocoumarin
PubChem CID 96929
IUPAC Name 7-(ethylamino)-4,6-dimethylchromen-2-one
SMILES CCNC1=C(C=C2C(=CC(=O)OC2=C1)C)C

Specifications

Melting Point 168.0°C to 170.0°C
Assay Percent Range 99%
Solubility Information Solubility in water: slightly soluble in water.
Infrared Spectrum Authentic
Lambda Max 366nm (in absolute ethanol)
Formula Weight 217.27
Percent Purity 99%
Grade Extra Pure
Molar Absorption Coefficient 20000L/(mole.cm) (at lambda min.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
UV_Vis 98.5% min.
Color Green/Yellow to Yellow
Quantity 500 mg
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Coumarin 2, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.