Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin-3-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. C28305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28305G 5 g
C28301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28305G Supplier TCI America Supplier No. C28305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15119-34-3
Molecular Formula C10H5NO2
Molecular Weight (g/mol) 171.16
MDL Number MFCD00115699
InChI Key QKJALQPLNMEDAV-UHFFFAOYSA-N
Synonym 3-cyanocoumarin, 2-oxo-2h-chromene-3-carbonitrile, coumarin-3-carbonitrile, 2h-1-benzopyran-3-carbonitrile, 2-oxo, 2-oxo-2h-1-benzopyran-3-carbonitrile, coumarin, 3-cyano-7ci,8ci, 3-cyano-coumarin, pubchem8675, acmc-1cfdq
PubChem CID 203763
IUPAC Name 2-oxo-2H-chromene-3-carbonitrile
SMILES O=C1OC2=CC=CC=C2C=C1C#N

Specifications

Melting Point 186°C
Color Yellow
Quantity 5 g
Formula Weight 171.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin-3-carbonitrile
RTECSNumber DJ2218100
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.