Learn More
Medchemexpress LLC HY-W027968 100mg Medchemexpress, Coumarin-3-carboxylic Acid CAS:531-81-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW027968100MG
Encompass_Preferred
Medchemexpress, HY-W027968 100mg Coumarin-3-carboxylic Acid CAS:531-81-7 Coumarin-3-carboxylic acid (2- Oxochromene-3-carboxylic acid) is an important initial compound for the synthesis of coumarins which are well known natural products for their diverse biological activities. Lanthanide complexes of Coumarin-3-carboxylic acid exhibit antiproliferative activity towards K-562 cell line. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0738