Learn More
Apexbio Technology LLC Coumarin-3-carboxylic Acid 531-81-7 100mg
SDP
Catalog No. 5000094637
Encompass_Preferred
Coumarin-3-carboxylic Acid is a coumarin-derived small molecule commonly applied as an investigative tool in biomedical research focusing on signal pathway regulation It is utilized to modulate enzymatic activities and cellular signaling pathways Coumarin-3-carboxylic Acid s biological activity has been studied in the context of its lanthanide complexes for inhibitory effects on cell growth in K-562 cell lines In cell-based studies it demonstrates antiproliferative activity in K-562 cells Based on these pharmacological properties Coumarin-3-carboxylic Acid holds research potential in elucidating biological mechanisms underlying hematologic malignancies and related diseases
Catalog No. 50-000-94637 Supplier Apexbio Technology LLC Supplier No. C7189100
Coumarin-3-carboxylic Acid is a coumarin-derived small molecule commonly applied as an investigative tool in biomedical research focusing on signal pathway regulation It is utilized to modulate enzymatic activities and cellular signaling pathways Coumarin-3-carboxylic Acid s biological activity has been studied in the context of its lanthanide complexes for inhibitory effects on cell growth in K-562 cell lines In cell-based studies it demonstrates antiproliferative activity in K-562 cells Based on these pharmacological properties Coumarin-3-carboxylic Acid holds research potential in elucidating biological mechanisms underlying hematologic malignancies and related diseases