Learn More
Medchemexpress LLC Coumarin-3-carboxylic acid | 531-81-7 | MFCD00006852 | 99.9% | 190.15 g/mol | C10H6O4 | 100 G
SDP
Catalog No. 5000301709
Encompass_Preferred
Coumarin-3-carboxylic acid is a coumarin derivative provided for research use as a white to off-white solid. It is used as a fluorescent probe for hydroxyl radicals and as a synthetic intermediate in coumarin chemistry. The material is offered in multiple package sizes to support analytical and preparative work.
  • High purity suitable for research applications.
  • Fluorescent probe for hydroxyl radical detection.
  • Available in multiple package sizes including 100 g for scale-up.
  • Solid white to off-white appearance, easy to weigh and store.
  • Well-characterized chemical identifiers: CAS 531-81-7, C10H6O4.
Catalog No. 50-003-01709 Supplier Medchemexpress LLC Supplier No. HYW027968100G
Coumarin-3-carboxylic acid is a coumarin derivative provided for research use as a white to off-white solid. It is used as a fluorescent probe for hydroxyl radicals and as a synthetic intermediate in coumarin chemistry. The material is offered in multiple package sizes to support analytical and preparative work.
  • High purity suitable for research applications.
  • Fluorescent probe for hydroxyl radical detection.
  • Available in multiple package sizes including 100 g for scale-up.
  • Solid white to off-white appearance, easy to weigh and store.
  • Well-characterized chemical identifiers: CAS 531-81-7, C10H6O4.