Learn More
Medchemexpress LLC Coumarin-3-carboxylic acid | 531-81-7 | MFCD00006852 | 99.9% | 190.15 g/mol | C10H6O4 | 25 G
SDP
Catalog No. 5000303360
Encompass_Preferred
Coumarin-3-carboxylic acid is a small-molecule fluorescent probe (C10H6O4, MW 190.15 g/mol) used to detect hydroxyl radicals and study oxidative stress in analytical and biochemical research. It is provided in high-purity solid form and as a DMSO solution for convenient use in assays and photophysical measurements.
  • High purity suitable for analytical and preparative work.
  • Strong fluorescence signal for sensitive radical detection.
  • Available in multiple package sizes and solution format.
  • Documented spectral and chromatographic data for quality assurance.
  • Applicable to in vitro assays and photophysical studies.
Catalog No. 50-003-03360 Supplier Medchemexpress LLC Supplier No. 25GHYW027968
Coumarin-3-carboxylic acid is a small-molecule fluorescent probe (C10H6O4, MW 190.15 g/mol) used to detect hydroxyl radicals and study oxidative stress in analytical and biochemical research. It is provided in high-purity solid form and as a DMSO solution for convenient use in assays and photophysical measurements.
  • High purity suitable for analytical and preparative work.
  • Strong fluorescence signal for sensitive radical detection.
  • Available in multiple package sizes and solution format.
  • Documented spectral and chromatographic data for quality assurance.
  • Applicable to in vitro assays and photophysical studies.