Learn More
Medchemexpress LLC Coumarin-3-carboxylic acid | 531-81-7 | MFCD00006852 | 99.9% | 190.15 g/mol | C10H6O4 | 500 G
SDP
Catalog No. 5000300231
Encompass_Preferred
Coumarin-3-carboxylic acid is an aromatic coumarin derivative (C10H6O4, MW 190.15 g/mol) commonly used as a fluorescence probe for detection of hydroxyl radicals and as a synthetic intermediate in organic and medicinal chemistry. It is supplied in high purity suitable for research and is available in a range of laboratory pack sizes.
  • Used as a fluorescence probe for hydroxyl radical detection
  • Serves as a synthetic intermediate for coumarin derivatives
  • High purity suitable for analytical and research use
  • Available in multiple laboratory pack sizes, including bulk options
  • Compatible with standard organic solvents for synthesis and analysis
Catalog No. 50-003-00231 Supplier Medchemexpress LLC Supplier No. HYW027968500G
Coumarin-3-carboxylic acid is an aromatic coumarin derivative (C10H6O4, MW 190.15 g/mol) commonly used as a fluorescence probe for detection of hydroxyl radicals and as a synthetic intermediate in organic and medicinal chemistry. It is supplied in high purity suitable for research and is available in a range of laboratory pack sizes.
  • Used as a fluorescence probe for hydroxyl radical detection
  • Serves as a synthetic intermediate for coumarin derivatives
  • High purity suitable for analytical and research use
  • Available in multiple laboratory pack sizes, including bulk options
  • Compatible with standard organic solvents for synthesis and analysis