Learn More
Medchemexpress LLC Coumarin 314 | 55804-66-5 | 250mg
SDP
Catalog No. 5000302741
Encompass_Preferred
Coumarin 314 is a dye which has an intense absorption in the visible and additionally presents a large solvent dependence[1]
Catalog No. 50-003-02741 Supplier Medchemexpress LLC Supplier No. 250MGHYW027857
Coumarin 314 is a dye which has an intense absorption in the visible and additionally presents a large solvent dependence[1]