Learn More
Medchemexpress LLC Coumarin 314 | 55804-66-5 | MFCD00051334 | 98.0% | 313.3 g/mol | C18H19NO4 | 500 MG
SDP
Catalog No. 5000301563
Encompass_Preferred
Coumarin 314 is a fluorescent dye that exhibits intense absorption in the visible region and pronounced solvent-dependent spectral behavior. It is used as a visible-absorbing fluorophore for photophysical studies, fluorescence labeling, and materials research, and is supplied with analytical documentation for research use.
  • Intense absorption in the visible region.
  • Pronounced solvent-dependent spectral behavior.
  • High stated purity of 98.0%.
  • Available in small research quantities (100 mg, 250 mg, 500 mg; larger sizes by request).
  • Manufacturer provides SDS, COA, and a data sheet for each batch.
  • Suitable for fluorescence labeling and photophysical studies.
Catalog No. 50-003-01563 Supplier Medchemexpress LLC Supplier No. HYW027857500MG
Coumarin 314 is a fluorescent dye that exhibits intense absorption in the visible region and pronounced solvent-dependent spectral behavior. It is used as a visible-absorbing fluorophore for photophysical studies, fluorescence labeling, and materials research, and is supplied with analytical documentation for research use.
  • Intense absorption in the visible region.
  • Pronounced solvent-dependent spectral behavior.
  • High stated purity of 98.0%.
  • Available in small research quantities (100 mg, 250 mg, 500 mg; larger sizes by request).
  • Manufacturer provides SDS, COA, and a data sheet for each batch.
  • Suitable for fluorescence labeling and photophysical studies.