Learn More
Medchemexpress LLC Coumarin 314 | 55804-66-5 | MFCD00051334 | 98.0% | 313.35 g·mol⁻¹ | C18H19NO4 | 100 MG
SDP
Catalog No. 5000300141
Encompass_Preferred
Coumarin 314 is a fluorescent dye exhibiting strong absorption in the visible region and pronounced solvent-dependent spectral properties. It is supplied as a light yellow solid and is used primarily as a research reagent in fluorescence measurements, probe design, and photophysical studies.
  • Strong visible absorption and fluorescence.
  • Pronounced solvent-dependent spectral behavior.
  • High purity for research applications.
  • Light yellow solid appearance for easy handling and identification.
  • Suitable for photophysical and fluorescence probe studies.
Catalog No. 50-003-00141 Supplier Medchemexpress LLC Supplier No. HYW027857100MG
Coumarin 314 is a fluorescent dye exhibiting strong absorption in the visible region and pronounced solvent-dependent spectral properties. It is supplied as a light yellow solid and is used primarily as a research reagent in fluorescence measurements, probe design, and photophysical studies.
  • Strong visible absorption and fluorescence.
  • Pronounced solvent-dependent spectral behavior.
  • High purity for research applications.
  • Light yellow solid appearance for easy handling and identification.
  • Suitable for photophysical and fluorescence probe studies.