Learn More
AdipoGen Coumarin 314
SDP
Catalog No. 502561491
Encompass_Preferred
Chemical. CAS 55804-66-5. Formula C18H19NO4. Molecular Weight 313.35. Coumarin 314 is a rigidized aminocoumarin derivative used as a laser dye with an absorption maximum of 436nm. It can be used as starting material for the synthesis of fluorescent probes. Spectral data lambdamax 436nm in ethanol, epsilon extinction coefficient >39000 at 389-401 nm in ethanol, lambdalaser 504nm.
Catalog No. 50-256-1491 Supplier AdipoGen Supplier No. CDXC0076G001
Chemical. CAS 55804-66-5. Formula C18H19NO4. Molecular Weight 313.35. Coumarin 314 is a rigidized aminocoumarin derivative used as a laser dye with an absorption maximum of 436nm. It can be used as starting material for the synthesis of fluorescent probes. Spectral data lambdamax 436nm in ethanol, epsilon extinction coefficient >39000 at 389-401 nm in ethanol, lambdalaser 504nm.