Learn More
AdipoGen Coumarin 334
SDP
Catalog No. 502561493
Encompass_Preferred
Chemical. CAS 55804-67-6. Formula C17H17NO3. Molecular Weight 283.32. Coumarin 334 is a laser dye with an absorption maximum of 452nm. It is used as starting material for the synthesis of cysteine-selective or pH indicator fluorescent probes. Spectral data lambdamax 452nm, epsilon extinction coefficient >45000 at 449-455 nm in ethanol, lambdalaser 532nm.
Catalog No. 50-256-1493 Supplier AdipoGen Supplier No. CDXC0075G001
Chemical. CAS 55804-67-6. Formula C17H17NO3. Molecular Weight 283.32. Coumarin 334 is a laser dye with an absorption maximum of 452nm. It is used as starting material for the synthesis of cysteine-selective or pH indicator fluorescent probes. Spectral data lambdamax 452nm, epsilon extinction coefficient >45000 at 449-455 nm in ethanol, lambdalaser 532nm.