Learn More
Cayman Chemical Coumarin 343, Purity: ≥95%, CAS: 55804-65-4, 250 mg
SDP
Catalog No. 502978423
Encompass_Preferred
A coumarin laser dye; abs/em = 446/519 nm
Catalog No. 50-297-8423 Supplier Cayman Chemical Supplier No. 41970250 MG
A coumarin laser dye; abs/em = 446/519 nm