Learn More
Medchemexpress LLC Coumarin 343 | 55804-65-4 | MFCD00051335 | 95.7% | 285.30 g/mol | C16H15NO4 | 50 MG
SDP
Catalog No. 5000282242
Encompass_Preferred
Coumarin 343 (C343) is a hydrophilic fluorescent probe for micro water pools, microfluidic systems, and fluorescence-based assays. Supplied as a solid dye with high purity, it is suitable for analytical measurements and fluorescent labeling. The dye exhibits blue emission with a peak near 425 nm and measurable emission across the 425-550 nm range.
  • Hydrophilic fluorescent probe for aqueous microenvironments.
  • Emission peak near 425 nm for blue fluorescence detection.
  • Emission range 425-550 nm provides a broad detection window.
  • High purity (95.7%) suitable for analytical and labeling work.
  • Supplied as a solid in small laboratory quantities for convenient handling.
Catalog No. 50-002-82242 Supplier Medchemexpress LLC Supplier No. HYW26736450MG
Coumarin 343 (C343) is a hydrophilic fluorescent probe for micro water pools, microfluidic systems, and fluorescence-based assays. Supplied as a solid dye with high purity, it is suitable for analytical measurements and fluorescent labeling. The dye exhibits blue emission with a peak near 425 nm and measurable emission across the 425-550 nm range.
  • Hydrophilic fluorescent probe for aqueous microenvironments.
  • Emission peak near 425 nm for blue fluorescence detection.
  • Emission range 425-550 nm provides a broad detection window.
  • High purity (95.7%) suitable for analytical and labeling work.
  • Supplied as a solid in small laboratory quantities for convenient handling.