Learn More
Medchemexpress LLC Coumarin 343 (C343) | 55804-65-4 | MFCD00051335 | 95.7% | 285.30 g/mol | C16H15NO4 | 100 MG
SDP
Catalog No. 5000280688
Encompass_Preferred
Coumarin 343 (C343) is a hydrophilic, blue-emitting coumarin dye used as a fluorescent probe to characterize micro water pools and microenvironments in fluorescence spectroscopy and imaging. It exhibits a pronounced emission in the blue region and is supplied as a research-grade reagent with documented solubility and analytical data.
Catalog No. 50-002-80688 Supplier Medchemexpress LLC Supplier No. HYW267364100MG
Coumarin 343 (C343) is a hydrophilic, blue-emitting coumarin dye used as a fluorescent probe to characterize micro water pools and microenvironments in fluorescence spectroscopy and imaging. It exhibits a pronounced emission in the blue region and is supplied as a research-grade reagent with documented solubility and analytical data.