Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coumarin 480 | 41267-76-9 | MFCD00041844 | 100.0% | 255.31 g/mol | C16H17NO2 | 100 MG
SDP

Catalog No. 5000282011
Encompass_Preferred

Coumarin 480 (Coumarin 102) is a laser dye and fluorescent probe used in photophysics and spectroscopy applications. Supplied as a high-purity solid for research use, the compound is typically used in fluorescence studies, solvent relaxation dynamics, and as a laser dye in optical experiments. Store protected from light and follow supplier documentation for handling and stability.

  • High purity: 99.95%.
  • Molecular weight: 255.31 g/mol.
  • Chemical formula: C16H17NO2.
  • Available in small-mass quantities for laboratory use.
  • Storage: keep at 4°C and protect from light.
  • In solvent stability: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-82011 Supplier Medchemexpress LLC Supplier No. HYW268197100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coumarin 480 (Coumarin 102) is a laser dye and fluorescent probe used in photophysics and spectroscopy applications. Supplied as a high-purity solid for research use, the compound is typically used in fluorescence studies, solvent relaxation dynamics, and as a laser dye in optical experiments. Store protected from light and follow supplier documentation for handling and stability.

  • High purity: 99.95%.
  • Molecular weight: 255.31 g/mol.
  • Chemical formula: C16H17NO2.
  • Available in small-mass quantities for laboratory use.
  • Storage: keep at 4°C and protect from light.
  • In solvent stability: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.