Learn More
Medchemexpress LLC Coumarin 480 | 41267-76-9 | MFCD00041844 | 100.0% | 255.31 g/mol | C16H17NO2 | 25 MG
SDP
Catalog No. 5000281410
Encompass_Preferred
Coumarin 480 (Coumarin 102) is a high-purity fluorescent laser dye used in fluorescence spectroscopy, laser applications, and optical research. It shows strong emission near 480 nm and is supplied as a research-grade reagent for imaging, sensor development, and photophysical studies. Users should follow the product SDS for safe handling and storage.
  • High purity (99.95%).
  • Strong fluorescence emission near 480 nm.
  • Suitable for fluorescence spectroscopy and laser dye applications.
  • Useful for sensor development and imaging studies.
  • Provided as a research reagent in small vials.
Catalog No. 50-002-81410 Supplier Medchemexpress LLC Supplier No. HYW26819725MG
Coumarin 480 (Coumarin 102) is a high-purity fluorescent laser dye used in fluorescence spectroscopy, laser applications, and optical research. It shows strong emission near 480 nm and is supplied as a research-grade reagent for imaging, sensor development, and photophysical studies. Users should follow the product SDS for safe handling and storage.
  • High purity (99.95%).
  • Strong fluorescence emission near 480 nm.
  • Suitable for fluorescence spectroscopy and laser dye applications.
  • Useful for sensor development and imaging studies.
  • Provided as a research reagent in small vials.