Learn More
Medchemexpress LLC Coumarin 480 (Coumarin 102) | 41267-76-9 | MFCD00041844 | 100.0% | 255.31 g·mol⁻¹ | C16H17NO2 | 50 MG
SDP
Catalog No. 5000281303
Encompass_Preferred
Coumarin 480 is a fluorescent coumarin-class laser dye supplied as a high-purity research reagent. It is intended for photophysical studies, fluorescent labeling, and laser applications. Certificate of analysis and safety data sheet are available from the manufacturer.
  • Laser dye suitable for photophysical and fluorescence studies.
  • High purity: 99.95%.
  • Solid form supplied in small laboratory pack sizes (25 mg, 50 mg, 100 mg; larger sizes available on request).
  • Documented safety and quality data available (COA and SDS).
  • Molecular formula C16H17NO2; molecular weight 255.31 g·mol⁻¹.
Catalog No. 50-002-81303 Supplier Medchemexpress LLC Supplier No. HYW26819750MG
Coumarin 480 is a fluorescent coumarin-class laser dye supplied as a high-purity research reagent. It is intended for photophysical studies, fluorescent labeling, and laser applications. Certificate of analysis and safety data sheet are available from the manufacturer.
  • Laser dye suitable for photophysical and fluorescence studies.
  • High purity: 99.95%.
  • Solid form supplied in small laboratory pack sizes (25 mg, 50 mg, 100 mg; larger sizes available on request).
  • Documented safety and quality data available (COA and SDS).
  • Molecular formula C16H17NO2; molecular weight 255.31 g·mol⁻¹.