Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 498 98.0+%, TCI America™
SDP

Catalog No. C3044200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3044200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3044200MG Supplier TCI America Supplier No. C3044200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87331-48-4
Molecular Formula C16H17NO4S
Molecular Weight (g/mol) 319.38
MDL Number MFCD00467041
InChI Key DPJWIXMOCGJKDG-UHFFFAOYSA-N
Synonym 2,3,6,7-Tetrahydro-10-(methylsulfonyl)-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one
PubChem CID 91972096
IUPAC Name 5-methanesulfonyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
SMILES CS(=O)(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O

Specifications

Melting Point 193°C
Quantity 200 mg
Formula Weight 319.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 498
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.