Learn More
Selleck Chemical LLC Coumarin 6 S5170-25mg
SDP
Catalog No. 503108226
Encompass_Preferred
Coumarin 6 is a fluorescent dye frequently used to facilitate the traceability of drug delivery systems in vitro
Catalog No. 50-310-8226 Supplier Selleck Chemical LLC Supplier No. S517025MG
Coumarin 6 is a fluorescent dye frequently used to facilitate the traceability of drug delivery systems in vitro