Learn More
Medchemexpress LLC Coumarin 6 | 38215-36-0 | 1 ML
SDP
Catalog No. 5000418132
Encompass_Preferred
Coumarin 6 is a fluorescent dye utilized as a fluorescent probe within microparticle drug delivery systems. It is employed to conduct in vivo tracking, cellular uptake studies, and investigations into the transport mechanisms of drug delivery systems. It has an excitation wavelength (λexc) of 450 nm and an emission wavelength (λem) of 505 nm. This product is intended for research use only and is not sold to patients.
  • Used for in vivo tracking, cell uptake, and transport mechanism studies in drug delivery systems
  • Optimal wavelengths: Excitation at 450 nm and emission at 505 nm
  • Purity: 99.59%
Catalog No. 50-004-18132 Supplier Medchemexpress LLC Supplier No. HYN713110MM/1ML
Coumarin 6 is a fluorescent dye utilized as a fluorescent probe within microparticle drug delivery systems. It is employed to conduct in vivo tracking, cellular uptake studies, and investigations into the transport mechanisms of drug delivery systems. It has an excitation wavelength (λexc) of 450 nm and an emission wavelength (λem) of 505 nm. This product is intended for research use only and is not sold to patients.
  • Used for in vivo tracking, cell uptake, and transport mechanism studies in drug delivery systems
  • Optimal wavelengths: Excitation at 450 nm and emission at 505 nm
  • Purity: 99.59%