Learn More
Medchemexpress LLC Coumarin 6 | 38215-36-0 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN7131100MG
Encompass_Preferred
Coumarin 6 is a fluorescent dye used as a fluorescent probe within microparticle drug delivery systems. It is designed for research use only and not sold to patients.
  • Fluorescent dye for microparticle drug delivery systems
  • Used for in vivo tracking
  • Supports cell uptake studies
  • Aids research into transport mechanisms of drug delivery systems
  • Excitation wavelength: 450 nm
  • Emission wavelength: 505 nm
Catalog No. 50-004-19297