Learn More
Apexbio Technology LLC Coumarin 6 38215-36-0 100mg
SDP
Catalog No. 5000094764
Encompass_Preferred
Coumarin 6 (CAS 38215-36-0) is a fluorescent dye utilized primarily as a tracer in biomedical research for particulate drug delivery systems Coumarin 6 facilitates the visualization and analysis of particle distribution trafficking and cellular uptake processes in vitro and in vivo It is applied to study biological mechanisms related to transport and localization of therapeutic molecules and their interactions with cellular compartments Based on these properties Coumarin 6 holds research potential in evaluating drug delivery dynamics cellular internalization pathways pharmacokinetic behavior and disease-related investigations
Catalog No. 50-000-94764 Supplier Apexbio Technology LLC Supplier No. A4770100
Coumarin 6 (CAS 38215-36-0) is a fluorescent dye utilized primarily as a tracer in biomedical research for particulate drug delivery systems Coumarin 6 facilitates the visualization and analysis of particle distribution trafficking and cellular uptake processes in vitro and in vivo It is applied to study biological mechanisms related to transport and localization of therapeutic molecules and their interactions with cellular compartments Based on these properties Coumarin 6 holds research potential in evaluating drug delivery dynamics cellular internalization pathways pharmacokinetic behavior and disease-related investigations