Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coumarin 6 | 38215-36-0 | 50 MG
SDP

Catalog No. 5000417193
Encompass_Preferred

Coumarin 6 is a fluorescent dye primarily employed as a probe within microparticle drug delivery systems. It facilitates in vivo tracking, cell uptake, and transport mechanism studies, with excitation at 450 nm and emission at 505 nm. This product is intended strictly for research purposes.

  • Fluorescent dye
  • Purity: 99.59%
  • Appearance: Solid
  • Color: Yellow to orange
  • Emission: 505
  • Excitation: 450

Catalog No. 50-004-17193 Supplier Medchemexpress LLC Supplier No. HYN713150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coumarin 6 is a fluorescent dye primarily employed as a probe within microparticle drug delivery systems. It facilitates in vivo tracking, cell uptake, and transport mechanism studies, with excitation at 450 nm and emission at 505 nm. This product is intended strictly for research purposes.

  • Fluorescent dye
  • Purity: 99.59%
  • Appearance: Solid
  • Color: Yellow to orange
  • Emission: 505
  • Excitation: 450

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.