Learn More
AdipoGen Coumarin-6-sulfonyl chloride
SDP
Catalog No. 502561537
Encompass_Preferred
Chemical. CAS 10543-42-7. Formula C9H5ClO4S. MW 244.7. Synthetic. Derivatizing agent for amines and phenols. Fluorescence is produced in alkaline conditions. Spectral properties lambdaex 360nm lambdaem 460nm in alkaline conditions.
Catalog No. 50-256-1537 Supplier AdipoGen Supplier No. CDXC0029M050
Chemical. CAS 10543-42-7. Formula C9H5ClO4S. MW 244.7. Synthetic. Derivatizing agent for amines and phenols. Fluorescence is produced in alkaline conditions. Spectral properties lambdaex 360nm lambdaem 460nm in alkaline conditions.