Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coumarin 6H | 58336-35-9 | 99.6% | 1 ML
SDP

Catalog No. 5000233710
Encompass_Preferred

Coumarin 6H is a coumarin-family laser dye supplied as a high-purity solid or as a 10 mM solution in dimethyl sulfoxide (DMSO). It exhibits strong fluorescence that can be enhanced through hydrogen-bonding interactions, making it suitable for laser dye and fluorescence applications.

  • Available as 10 mM solution in DMSO or as solid quantities (50 mg-1 g).
  • High purity, typically 99.6%.
  • Molecular formula C15H15NO2 and molecular weight 241.29 g·mol^-1.
  • Fluorescence emission enhanced by hydrogen bonding interactions.
  • Soluble in common organic solvents such as DMSO.
  • Intended for use as a laser dye and fluorescent probe in optical applications.

Catalog No. 50-002-33710 Supplier Medchemexpress LLC Supplier No. HY12193010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coumarin 6H is a coumarin-family laser dye supplied as a high-purity solid or as a 10 mM solution in dimethyl sulfoxide (DMSO). It exhibits strong fluorescence that can be enhanced through hydrogen-bonding interactions, making it suitable for laser dye and fluorescence applications.

  • Available as 10 mM solution in DMSO or as solid quantities (50 mg-1 g).
  • High purity, typically 99.6%.
  • Molecular formula C15H15NO2 and molecular weight 241.29 g·mol^-1.
  • Fluorescence emission enhanced by hydrogen bonding interactions.
  • Soluble in common organic solvents such as DMSO.
  • Intended for use as a laser dye and fluorescent probe in optical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.