Learn More
Medchemexpress LLC Coumarin 6H | 58336-35-9 | 99.6% | 250 MG
SDP
Catalog No. 5000232918
Encompass_Preferred
Coumarin 6H is a coumarin-derived laser dye that functions as a fluorescent probe for spectroscopy and imaging applications. It exhibits enhanced fluorescence through hydrogen-bonding interactions, and is supplied as a solid powder for laboratory use.
  • High purity fluorescent dye for analytical applications.
  • Solid powder, yellow to brown appearance for easy handling and storage.
  • Enhanced fluorescence emission via hydrogen bonding interactions.
  • Available in small-scale package sizes suitable for research use.
  • Store protected from light at low temperatures to maintain stability.
Catalog No. 50-002-32918 Supplier Medchemexpress LLC Supplier No. HY121930250MG
Coumarin 6H is a coumarin-derived laser dye that functions as a fluorescent probe for spectroscopy and imaging applications. It exhibits enhanced fluorescence through hydrogen-bonding interactions, and is supplied as a solid powder for laboratory use.
  • High purity fluorescent dye for analytical applications.
  • Solid powder, yellow to brown appearance for easy handling and storage.
  • Enhanced fluorescence emission via hydrogen bonding interactions.
  • Available in small-scale package sizes suitable for research use.
  • Store protected from light at low temperatures to maintain stability.