Learn More
Medchemexpress LLC Coumarin 7 | 27425-55-4 | MFCD00051348 | 99.8% | 333.4 g/mol | C20H19N3O2 | 50MG
SDP
Catalog No. 5000238014
Encompass_Preferred
Coumarin 7 is a coumarin-class laser dye supplied as a high-purity solid for research applications. It is used as a fluorescent probe and laser medium in spectroscopy, fluorescence studies, and related dye research, and is provided in small, laboratory-scale quantities. Intended for research use only.
  • Laser-active coumarin dye with strong fluorescence.
  • High purity (99.8%) suitable for analytical and research applications.
  • Provided in mg-scale quantities for laboratory experiments.
  • Solid powder form, easy to weigh and dissolve in common organic solvents.
  • Intended for research use only.
Catalog No. 50-002-38014 Supplier Medchemexpress LLC Supplier No. HY12575050MG
Coumarin 7 is a coumarin-class laser dye supplied as a high-purity solid for research applications. It is used as a fluorescent probe and laser medium in spectroscopy, fluorescence studies, and related dye research, and is provided in small, laboratory-scale quantities. Intended for research use only.
  • Laser-active coumarin dye with strong fluorescence.
  • High purity (99.8%) suitable for analytical and research applications.
  • Provided in mg-scale quantities for laboratory experiments.
  • Solid powder form, easy to weigh and dissolve in common organic solvents.
  • Intended for research use only.