Learn More
AdipoGen Coumarin 7
SDP
Catalog No. 502561496
Encompass_Preferred
Chemical. CAS 27425-55-4. Formula C20H19N3O2. Molecular Weight 333.38. Coumarin 7 is a greenish yellow fluorescent dye. It has good heat resistance and light resistance, good migration resistance and high tinting strength with wide application. Coumarin 7 is a laser dye that is used as green phosphorescent dopant material for organic light-emitting diodes OLED. Its derivatives are used in medicine and chemistry. Spectral Data lambdaabs 438nm, lambdaem 266nm/355nm, lambdaex 490nm, lambdalaser 520nm.
Catalog No. 50-256-1496 Supplier AdipoGen Supplier No. CDXC0071G001
Chemical. CAS 27425-55-4. Formula C20H19N3O2. Molecular Weight 333.38. Coumarin 7 is a greenish yellow fluorescent dye. It has good heat resistance and light resistance, good migration resistance and high tinting strength with wide application. Coumarin 7 is a laser dye that is used as green phosphorescent dopant material for organic light-emitting diodes OLED. Its derivatives are used in medicine and chemistry. Spectral Data lambdaabs 438nm, lambdaem 266nm/355nm, lambdaex 490nm, lambdalaser 520nm.