Learn More
Medchemexpress LLC Coumarin | 91-64-5 | 146.15 | 5 G
SDP
Catalog No. 5000417002
Encompass_Preferred
Coumarin is a potent and orally active anti-inflammatory agent. It also demonstrates antinociceptive, antibacterial, antifungal, and anticancer activities.
  • Potent and orally active anti-inflammatory agent
  • Shows antinociceptive effect
  • Shows antibacterial, antifungal and anticancer activity
Catalog No. 50-004-17002 Supplier Medchemexpress LLC Supplier No. HYN07095G
Coumarin is a potent and orally active anti-inflammatory agent. It also demonstrates antinociceptive, antibacterial, antifungal, and anticancer activities.
  • Potent and orally active anti-inflammatory agent
  • Shows antinociceptive effect
  • Shows antibacterial, antifungal and anticancer activity