Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin, 98%

Catalog No. aaa1533630
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1533630 250 g
AAA153360B 1000 g
AAA153360I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1533630 Supplier Thermo Scientific Chemicals Supplier No. A1533630
Only null left
Add to Cart

CAS: 91-64-5 | C9H6O2 | 146.15 g/mol

Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-64-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006850
InChI Key ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
Synonym coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone
PubChem CID 323
ChEBI CHEBI:28794
SMILES O=C1OC2=CC=CC=C2C=C1

Specifications

Melting Point 68°C to 71°C
Density 0.935
Boiling Point 297°C to 299°C
Flash Point 162°C (323°F)
Odor Pleasant
Quantity 250 g
UN Number UN2811
Beilstein 383644
Merck Index 14,2562
Solubility Information Soluble in water.
IUPAC Name 2H-chromen-2-one
Formula Weight 146.15
Percent Purity 98%
Chemical Name or Material Coumarin
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H351
Toxic if swallowed.
Suspected of causing cancer.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-086-7
RTECSNumber GN4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.