Learn More
Medchemexpress LLC Coumarin boronic aci 10mM 1mL
SDP
Catalog No. 5000206588
Encompass_Preferred
Coumarin boronic acid is a fluorescent probe The excitation and emission wavelengths of coumarin boronic acid are set to 360 nm and 430 nm respectively Coumarin boronic acid can be used to monitor the formation of amino acid and protein hydroxyl peroxides in real time which is beneficial for understanding the mechanisms of oxidative stress and protein post-translational modification[1]
Catalog No. 50-002-06588 Supplier Medchemexpress LLC Supplier No. HYW11660610MM/1ML
Coumarin boronic acid is a fluorescent probe The excitation and emission wavelengths of coumarin boronic acid are set to 360 nm and 430 nm respectively Coumarin boronic acid can be used to monitor the formation of amino acid and protein hydroxyl peroxides in real time which is beneficial for understanding the mechanisms of oxidative stress and protein post-translational modification[1]