Learn More
Medchemexpress LLC Coumarin boronic aci 5mg
SDP
Catalog No. 5000206774
Encompass_Preferred
Coumarin boronic acid is a fluorescent probe The excitation and emission wavelengths of coumarin boronic acid are set to 360 nm and 430 nm respectively Coumarin boronic acid can be used to monitor the formation of amino acid and protein hydroxyl peroxides in real time which is beneficial for understanding the mechanisms of oxidative stress and protein post-translational modification[1]
Catalog No. 50-002-06774 Supplier Medchemexpress LLC Supplier No. HYW1166065MG
Coumarin boronic acid is a fluorescent probe The excitation and emission wavelengths of coumarin boronic acid are set to 360 nm and 430 nm respectively Coumarin boronic acid can be used to monitor the formation of amino acid and protein hydroxyl peroxides in real time which is beneficial for understanding the mechanisms of oxidative stress and protein post-translational modification[1]