Learn More
Cayman Chemical Coumarin Boronic Acid, Purity: ≥98%, CAS: 1357078-03-5, 100 mg
SDP
Catalog No. 502953820
Encompass_Preferred
A fluorescent probe for direct detection of peroxynitrite, hypochlorous acid, and hydrogen peroxide; reacts with peroxynitrite at an exponentially faster rate (k = 1.1 μM/s) than hydrogen peroxide (k =1.5 M/s); COH displays ex/em = 332/470 nm, respectively
Catalog No. 50-295-3820 Supplier Cayman Chemical Supplier No. 14051100 MG
A fluorescent probe for direct detection of peroxynitrite, hypochlorous acid, and hydrogen peroxide; reacts with peroxynitrite at an exponentially faster rate (k = 1.1 μM/s) than hydrogen peroxide (k =1.5 M/s); COH displays ex/em = 332/470 nm, respectively