Learn More
Cayman Chemical Coumarin Boronic Acid, 1357078-03-5, 10 mg
SDP
Supplier:  Cayman Chemical 1405110
Encompass_Preferred
A fluorescent probe that reacts stoichiometrically and rapidly with ONOO- (rate constant (k) = 1.1 μM/s); excitation of 332 nm and emission of 470 nm
Catalog No. NC0766049
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.