Learn More
Cayman Chemical Coumarin Boronic Acid pinacolate ester, Purity: ≥98%, CAS: 190788-61-5, 10 mg
SDP
Catalog No. 502889755
Encompass_Preferred
A fluorescent probe for detection of peroxynitrite; a more soluble form of CBA; can be used to detect peroxynitrite, hypochlorous acid, and hydrogen peroxide; COH displays ex/em = 332/470 nm, respectively
Catalog No. 50-288-9755 Supplier Cayman Chemical Supplier No. 1081810 MG
A fluorescent probe for detection of peroxynitrite; a more soluble form of CBA; can be used to detect peroxynitrite, hypochlorous acid, and hydrogen peroxide; COH displays ex/em = 332/470 nm, respectively