Learn More
Medchemexpress LLC Coumarin-C2-exo-BCN | 2488174-17-8 | 95.7% | 479.57 g·mol⁻¹ | C27H33N3O5 | 10 MG
SDP
Catalog No. 5000287820
Encompass_Preferred
Coumarin-C2-exo-BCN is a coumarin-derived fluorescent dye functionalized with an exo-BCN strained alkyne for bioorthogonal labeling via strain-promoted alkyne-azide cycloaddition (SPAAC). It is intended for catalyst-free conjugation to azide-containing biomolecules to generate fluorescent triazole-linked products.
  • Fluorescent coumarin scaffold for sensitive detection.
  • exo-BCN strained alkyne enables catalyst-free SPAAC with azides.
  • Used to prepare fluorescent conjugates and label azide-containing biomolecules.
  • Supplied as a solid with reported purity of ~95.7%.
Catalog No. 50-002-87820 Supplier Medchemexpress LLC Supplier No. HY15630510MG
Coumarin-C2-exo-BCN is a coumarin-derived fluorescent dye functionalized with an exo-BCN strained alkyne for bioorthogonal labeling via strain-promoted alkyne-azide cycloaddition (SPAAC). It is intended for catalyst-free conjugation to azide-containing biomolecules to generate fluorescent triazole-linked products.
  • Fluorescent coumarin scaffold for sensitive detection.
  • exo-BCN strained alkyne enables catalyst-free SPAAC with azides.
  • Used to prepare fluorescent conjugates and label azide-containing biomolecules.
  • Supplied as a solid with reported purity of ~95.7%.